• USA Home
  • D4524 - Dynorphin A Porcine Fragment 1-7

EMAIL THIS PAGE TO A FRIEND
D4524 Sigma-Aldrich

Dynorphin A Porcine Fragment 1-7

≥95% (HPLC)

  • CAS Number 77101-32-7

  • Empirical Formula (Hill Notation) C40H61N13O9

  • Molecular Weight 867.99

Purchase

Properties

assay   ≥95% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C

Description

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Leu-Arg-Arg

Biochem/physiol Actions

Potent non-selective opioid agonist

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?