EMAIL THIS PAGE TO A FRIEND
D6023 Sigma

2,4-Dichlorophenol

melt

  • CAS Number 120-83-2

  • Linear Formula Cl2C6H3OH

  • Molecular Weight 163.00

  •  Beilstein Registry Number 742467

  •  EC Number 204-429-6

  •  MDL number MFCD00002169

Purchase

Properties

InChI Key   HFZWRUODUSTPEG-UHFFFAOYSA-N
form   melt
bp   209-210 °C(lit.)
mp   42-43 °C (lit.)

Description

Physical form

Packaged as a melt

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2928 - class 6.1 - PG 2 - EHS - RQ - acidic - Toxic solids, corrosive, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
SK8575000
Flash Point(F) 
237.2 °F
Flash Point(C) 
114 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

105953 2,4-Dichlorophenol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?