• USA Home
  • D7438 - Monoclonal Anti-DP2 antibody produced in mouse

D7438 Sigma

Monoclonal Anti-DP2 antibody produced in mouse

~0.5 mg/mL, clone BC-2, purified immunoglobulin, buffered aqueous solution



Related Categories Alphabetical Index, Antibodies, Antibodies for Gene Regulation, Antibodies to Common Transcription Factors, Antibodies to Transcription Factors,
species reactivity   human
application(s)   immunoprecipitation (IP): suitable using native and denatured DP2
  microarray: suitable
  western blot: 0.5-1 μg/mL using a whole extract of transfected 293T (human embryonal kidney) cells expressing DP2
clone   BC-2, monoclonal
concentration   ~0.5 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt 47 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... TFDP2(7029)
biological source   mouse
conjugate   unconjugated



recombinant human DP2 protein.


Does not detect the other members of the DP family. May also be used in a gel mobility shift assay.

Preparation Note

Purified from culture supernatant of hybridoma cells grown in a bioreactor.


Monoclonal Anti-DP2 antibody produced in mouse is suitable for immunoblot assay at a working concentration of 0.5 - 1 μg/mL using a whole extract of transfected 293T (human embryonal kidney) cells expressing DP2, microarray and immunoprecipitation using native and denatured DP2. It may be used for electrophoretic mobility shift assay (EMSA).

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

DPs or dimerization partners (DP1 and DP2) form heterodimers along with E2 transcription factors. These active E2F transcriptional complexes bind to and regulate the transcription of several genes involved in control of the cell cycle regulation and DNA replication. Antibodies reacting specifically with DP2 are useful for the study of control of transcription in intracellular pathways, and its essential roles during developmental and pathological processes.

Price and Availability

Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  2
Protocols & Articles


Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers


Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?