EMAIL THIS PAGE TO A FRIEND
D7566 Sigma-Aldrich

Dichloromethane

suitable for determination of keto-steroids

Synonym: Methylene chloride

  • CAS Number 75-09-2

  • Empirical Formula (Hill Notation) CH2Cl2

  • Molecular Weight 84.93

  •  Beilstein/REAXYS Number 1730800

  •  EC Number 200-838-9

  •  MDL number MFCD00000881

Purchase

Properties

vapor density   2.9 (vs air)
vapor pressure   24.45 psi ( 55 °C)
  6.83 psi ( 20 °C)
autoignition temp.   1223 °F
expl. lim.   22 %
refractive index   n20/D 1.424 (lit.)
bp   39.8-40 °C (lit.)
mp   −97 °C (lit.)
density   1.325 g/mL at 25 °C (lit.)
suitability   suitable for determination of keto-steroids
SMILES string   ClCCl
InChI   1S/CH2Cl2/c2-1-3/h1H2
InChI key   YMWUJEATGCHHMB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2
RTECS 
PA8050000
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Content

Carcinogen Selector - Digestive System - Liver

From our library of Related Content, Sigma-Aldrich presents Carcinogen Selector - Digestive System - Liver
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Liver Endocrine

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System - Mammary Gland

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Endocrine System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Reproductive System - Lung

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Carcinogen Selector - Respiratory System

The Carcinogen Selector gives researchers the ability to select carcinogens by the mouse organs that they promote cancer in. The tool includes over 750 carcinogens that target 35 specific organs acro...
Keywords: Cancer, Carcinogens, Respiratory

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?