EMAIL THIS PAGE TO A FRIEND
D9648 Sigma

DIPSO

≥99% (titration)

Synonym: 3-(N,N-Bis[2-hydroxyethyl]amino)-2-hydroxypropanesulfonic acid, N,N-Bis(2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid

  • CAS Number 68399-80-4

  • Empirical Formula (Hill Notation) C7H17NO6S

  • Molecular Weight 243.28

  •  Beilstein Registry Number 2105454

  •  EC Number 269-992-2

  •  MDL number MFCD00038353

Purchase

Properties

InChI Key   XCBLFURAFHFFJF-UHFFFAOYSA-N
assay   ≥99% (titration)
useful pH range   7.0 - 8.2
pKa (25 °C)   7.6
mp   189-192 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Buffer Reference Center

Citric Acid – Na2HPO4 Buffer Solutions, pH approx. 2.6–7.61 Citric acid monohydrate, C6H8O7 • H2O, M. wt. 210.14; 0.1M-solution contains 21.01 g/l. Na2HPO4, M. wt. 141.98; 0.2M-solution contains 28.4...
Keywords: Biological Buffers, Buffers

缓冲液参考中心

柠檬酸 – Na2HPO4 缓冲液,pH约为 2.6–7.61 柠檬酸一水合物, C6H8O7 • H2O, M. wt. 210.14; 0.1M的溶液中含有21.01 g/l。 Na2HPO4, M. wt. 141.98; 0.2M的溶液中含有28.40g / l 或 Na2HPO4 • 2H2O, M. wt. 178.05; 0.2M的溶液中含有35.61 g/l。 xml 0.1M的...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?