• USA Home
  • D9783 - [D-Arg8]-Dynorphin A Porcine fragment 1-13

EMAIL THIS PAGE TO A FRIEND
D9783 Sigma-Aldrich

[D-Arg8]-Dynorphin A Porcine fragment 1-13

≥97% (HPLC)

  • CAS Number 125357-12-2

  • Empirical Formula (Hill Notation) C75H127N27O15

  • Molecular Weight 1646.98

  •  MDL number MFCD00076370

  •  PubChem Substance ID 24894331

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C
SMILES string   CC(C)CC(NC(=O)C(CCCCN)NC(=O)C1CCCN1C(=O)C(CCCNC(N)=N)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCNC(N)=N)NC(=O)C(CC(C)C)NC(=O)C(Cc2ccccc2)NC(=O)CNC(=O)CNC(=O)C(N)Cc3ccc(O)cc3)C(=O)NC(CCCCN)C(O)=O
Gene Information   pig ... PDYN(445529)

Description

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Leu-Arg-Arg-D-Arg-Arg-Pro-Lys-Leu-Lys

Biochem/physiol Actions

κ opioid receptor agonist

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?