• USA Home
  • DAX01 - AIM Biotech 3D Cell Culture Chips

EMAIL THIS PAGE TO A FRIEND
DAX01 Sigma-Aldrich

AIM Biotech 3D Cell Culture Chips

Synonym: Microfluidic chips

Purchase

Properties

material   plastic
sterility   sterile
packaging   box of 25 ea Individually packed sterile chips
availability   not available in South Korea, Singapore
storage temp.   room temp

Description

Application

An easy-to-use, modular platform to co-culture different cell types in discrete 3D and 2D compartments.
Applications include angiogenesis, vasculogenesis, metastasis (spheroid dispersion, intra- & extravasation), immuno-oncology (immune checkpoint, T-cell killing efficiency), 3D culture of tumor spheroids/ biopsies/ organoids, and other organotypic models.

Features and Benefits

• Microscope slide format 75 mm × 25 mm
• Compatible with all polymerizable gels including collagen, fibrinogen, Matrigel, etc. and combinations thereof
• Gas permeable laminate for effective gas exchange
• Optically clear and compatible with phase contrast, fluorescence and confocal microscopy
• Enables monotypic or organotypic co-culture models
• Enables the control of interstitial flow across the 3D gel region
• Enables the control of chemical gradients across the 3D gel region
• Sterile & ready-to-use
• Designed for rapid media exchange through vacuum aspiration with no risk of over-aspiration
• Designed for modular expansion with AIM Luer Connectors
• Fits into AIM Microtiter Plate Holders for easy handling and stacking

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Create a 3D kidney nephrotoxicity model on the AIM Biotech 3D cell culture chip

The SA7K cell line (MTOX1030) is a pseudo-immortalized human renal proximal tubule epithelial cell (RPTEC) line that was developed from primary cells using the CompoZr® Zinc Finger Nuclease technolog...
Keywords: Angiogenesis, Cell culture, Chromatin immunoprecipitation, Confocal microscopy, Drug discovery, Hormones, Immunofluorescence, Microscopy, Sample preparations

Human Renal Proximal Tubule Epithelial Cells (RPTEC) as an Essential Advanced Model to Express Kidney Drug Transporters & Toxicity

Human renal proximal tubule epithelial cells (RPTECs) are commonly used to predict human renal drug permeability and to investigate drug efflux. However, current methods for investigating drug-transp...
Keywords: Cell culture, Chromatin immunoprecipitation, Gene expression, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Polymerase chain reaction

Related Products

related product

Product #

Description

Add to Cart

HOL01 AIM Biotech 3D Cell Culture Chip Holders
LUC01 AIM Biotech 3D Cell Culture Chip Luer Connectors

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?