• USA Home
  • DOPA1050 - DOPA Neuronal Medium - 50mL

EMAIL THIS PAGE TO A FRIEND
DOPA1050 Sigma-Aldrich

DOPA Neuronal Medium - 50mL

Purchase

Properties

Related Categories Cell Biology, General Stem Cell Biology, Induced Pluripotent Stem Cells and Reagents, Media for iPSC-derived Differentiated Neural Cells, Stem Cell Biology,
sterility   sterile
form   liquid
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −20°C

Description

Preparation Note

See technical bulletin.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Neural Stem Cell Culture

Neural Stem Cell Lines Neural Stem Cell Media and Supplements Neural Stem Cell Markers Stem Cell Culture Protocols
Keywords: Cell biology, Cell culture, Central Nervous System, Cryopreservation, Diseases, Dopamine agents, Environmental, Gene expression, Growth factors, Neuroscience, Phase transitions, Stem cell biology, Transcription

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?