EMAIL THIS PAGE TO A FRIEND
E1139 Supelco

Estazolam solution

drug standard, 100 μg/L in methanol

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Benzodiazepines, Pharmaceuticals, Illicit Drugs & Alcohol
grade   drug standard
assay   ≥98%
drug control   Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   100 μg/L in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   2-8°C
SMILES string   Clc1ccc-2c(c1)C(=NCc3nncn-23)c4ccccc4
InChI   1S/C16H11ClN4/c17-12-6-7-14-13(8-12)16(11-4-2-1-3-5-11)18-9-15-20-19-10-21(14)15/h1-8,10H,9H2
InChI key   CDCHDCWJMGXXRH-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?