• USA Home
  • E4141 - Monoclonal Anti-S1P3, C-Terminal antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
E4141 Sigma-Aldrich

Monoclonal Anti-S1P3, C-Terminal antibody produced in mouse

clone AS61, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-EDG-3, Anti-Endothelial Cell Differentiation Gene 3, C-Terminal

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Lipids in Cell Signaling, Primary Antibodies,
conjugate   unconjugated
clone   AS61, monoclonal
biological source   mouse
application(s)   western blot: 1:10,000 using human and rat spleen and lung tissue
species reactivity   human, rat
mol wt   antigen 45 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG
Quality Level   200
antibody product type   primary antibodies
concentration   ~1 mg/mL
UniProt accession no.   Q99500
Gene Information   human ... S1PR3(1903)
rat ... Edg3l(29736)

Description

Immunogen

human EDG-3/S1P3 unique C-terminal peptide.

Application

Monoclonal Anti-S1P3, C-Terminal antibody produced in mouse is suitable for western blotting at a dilution of 1:10,000 using human and rat spleen and lung tissue.

Physical form

Solution in phosphate buffered saline with 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

Sphingosine-1-phosphate receptor 3 also known as S1PR3 is a human gene which encodes a G protein-coupled receptor. It binds the lipid signaling molecule sphingosine 1-phosphate (S1P). S1PR3 acts as a critical signaling molecule mediating cell proliferation and vascular permeability. It acts as a potential biomarker in murine and human acute lung injury (ALI). Sphingosine 1-phosphate (S1P) and the S1P receptor S1PR3 are intimately involved in lung inflammatory responses and vascular barrier regulation. S1PR3 serves as a biomarker of severity in critically acute respiratory distress syndrome (ARDS) patients and can act as a potential target for individualized therapy.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?