• USA Home
  • E4654 - Anti-EphB2 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
E4654 Sigma-Aldrich

Anti-EphB2 antibody produced in goat

affinity isolated antibody, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Angiogenesis, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology,
conjugate   unconjugated
clone   polyclonal
biological source   goat
application(s)   indirect ELISA: 0.5-1.0 μg/mL
  western blot: 0.1-0.2 μg/mL
species reactivity   mouse
form   lyophilized powder
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
UniProt accession no.   P54763
Gene Information   mouse ... Ephb2(13844)

Description

Specificity

In immunoblotting, the antibody exhibits ~5% cross-reactivity with recombinant rat EphB1 and EphA5, recombinant mouse EphA8, EphB6, EphA3, EphA4, EphA7, EphA2, EphB3, and recombinant human EphA1.

Immunogen

purified recombinant mouse EphB2 extracellular domain expressed in NSO cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?