• USA Home
  • E6636 - β-Endorphin human fragment 1-27

EMAIL THIS PAGE TO A FRIEND
E6636 Sigma-Aldrich

β-Endorphin human fragment 1-27

≥97% (HPLC)

  • CAS Number 76622-84-9

  • Empirical Formula (Hill Notation) C139H217N33O40S

  • Molecular Weight 3022.47

  •  MDL number MFCD00076389

  •  PubChem Substance ID 24894633

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Endorphins, Opioid Peptides, Peptides and Proteins,
InChI Key   CMLHEPBOBGYCBC-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... POMC(5443)

Description

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-Tyr

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?