• USA Home
  • EP0056000003 - Temperature Verification System

EMAIL THIS PAGE TO A FRIEND
EP0056000003

Temperature Verification System

Purchase

Properties

Related Categories Eppendorf, Eppendorf PCR, Eppendorf Thermal Cyclers and Accessories, Labware, Manufacturer Browser More...
feature   USB, single channel
  for use with Mastercycler® Pro
  verification meter
  without sensor
mfr. no.   Eppendorf®, 0056000003 (Global)
storage temp.   room temp

Description

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Mastercycler is a registered trademark of Eppendorf AG

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?