• USA Home
  • EPCO48220005 - Eppendorf® Galaxy® 48R CO2 Incubator, 48 L, 3 (6 optional) shelves

EMAIL THIS PAGE TO A FRIEND
EPCO48220005

Eppendorf® Galaxy® 48R CO2 Incubator, 48 L, 3 (6 optional) shelves

AC/DC input 120 V, parameter 1-19% (O2 control)

Purchase

Properties

AC/DC input   120 V
material   stainless steel
mfr. no.   Eppendorf®, CO48220005
parameter   1-19% (O2 control)
  4-50 °C temp. range

Description

General description

Stackable CO2 incubator with 48 L capacity, advanced model

Features and Benefits

• 48 Liter (1.7 ft3) provides mid-sized capacity within a small footprint
• Options: High Temp. Disinfection (4 hour at 120 °C), O2 control (e.g. for hypoxic studies), 2-split inner glass door
• Fanless design offers more usable space
• Six-sided direct heating system provides a uniform incubated environment
• Deep-drawn, easy-to-clean stainless steel chamber without seams or welds
• Low gas consumption
• Integrated alarm relay

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Galaxy is a registered trademark of Eppendorf AG

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?