EMAIL THIS PAGE TO A FRIEND
F0885 Sigma-Aldrich

Fmoc-Lys(Z)-OH

Synonym: Nα-Fmoc-Nε-Z-L-lysine

  • CAS Number 86060-82-4

  • Empirical Formula (Hill Notation) C29H30N2O6

  • Molecular Weight 502.56

  •  Beilstein/REAXYS Number 3578530

  •  MDL number MFCD00065662

  •  PubChem Substance ID 329799670

Purchase

Properties

InChI Key   KRULQRVJXQQPQH-SANMLTNESA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

47577 Fmoc-Lys(Z)-OH, ≥98.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?