EMAIL THIS PAGE TO A FRIEND
F3018 Sigma-Aldrich

Fetal Bovine Serum

Heat Inactivated, USA origin, sterile-filtered, suitable for insect cell culture

Synonym: FBS, FCS, sera, serum

Purchase

Properties

biological source   bovine fetus
sterility   sterile-filtered
composition   Hemoglobin, ≤20 mg/dL
origin   USA origin
application(s)   cell culture | insect: suitable
impurities   ≤10 EU/mL endotoxin
shipped in   dry ice
storage temp.   −20°C

Description

Application

Heat inactivated FBS is used in special cell culture applications where complement activity (normally present) would interfere with the cell culture objectives.

Linkage

Request a serum sample

Preparation Note

Prepared by heating at 56 °C for 30 minutes.

Analysis Note

• Endotoxin and hemoglobin tested

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

F4135 Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?