EMAIL THIS PAGE TO A FRIEND
F6387 Sigma-Aldrich

Fast Red GL Salt

Synonym: 4-Methyl-2-nitrobenzenediazonium 1,5-naphthalene disulfonate, Azoic diazo component 8

  • CAS Number 85223-03-6

  • Empirical Formula (Hill Notation) C17H13N3O8S2

  • Molecular Weight 451.43

  •  Colour Index Number 37110

  •  EC Number 286-355-4

  •  MDL number MFCD00057926

Purchase

Properties

composition   Dye content, ~90%
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?