• USA Home
  • F8501 - Formimino-L-aspartic acid

EMAIL THIS PAGE TO A FRIEND
F8501 Sigma-Aldrich

Formimino-L-aspartic acid

  • CAS Number 2374-41-6

  • Empirical Formula (Hill Notation) C5H8N2O4

  • Molecular Weight 160.13

  •  MDL number MFCD00056696

Purchase

Properties

storage temp.   −20°C
SMILES string   OC(=O)CC(NC=N)C(O)=O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?