• USA Home
  • F9134 - Flurazepam dihydrochloride

EMAIL THIS PAGE TO A FRIEND
F9134 Supelco

Flurazepam dihydrochloride

drug standard, 1.0 mg/mL in methanol

Synonym: 7-Chloro-1-(diethylamino)ethyl-5-(2-fluorophenyl)-3H-1,4-benzodiazepin-2(1H)-one dihydrochloride

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Benzodiazepines, Pharmaceuticals, Illicit Drugs & Alcohol
grade   drug standard
assay   ≥98%
drug control   Home Office Schedule 4.1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Forensics and Toxicology
Pharmaceutical (small molecule)
format   single component solution
storage temp.   2-8°C
SMILES string   Cl[H].Cl[H].CCN(CC)CCN1C(=O)CN=C(c2ccccc2F)c3cc(Cl)ccc13
InChI   1S/C21H23ClFN3O.2ClH/c1-3-25(4-2)11-12-26-19-10-9-15(22)13-17(19)21(24-14-20(26)27)16-7-5-6-8-18(16)23;;/h5-10,13H,3-4,11-12,14H2,1-2H3;2*1H
InChI key   ZIIJJOPLRSCQNX-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Benzodiazepine anxiolytic; anticonvulsant; ligand for the GABAA receptor benzodiazepine modulatory site.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?