• USA Home
  • F9800 - Anti-FLIP, N-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
F9800 Sigma-Aldrich

Anti-FLIP, N-Terminal antibody produced in rabbit

1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-CASH, Anti-CLARP, Anti-Casper, Anti-FLAME-1, Anti-I-FLICE

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Suppressors of Apoptosis,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 0.5-1 μg/mL
species reactivity   mouse, human, rat
mol wt   antigen 25 kDa (FLIPδ/FLIPS)
  antigen 35 kDa (FLIPγ)
  antigen 52 kDa (FLIPβ)
  antigen 55 kDa (FLIPα/FLIPL)
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   1 mg/mL
UniProt accession no.   O15519
Gene Information   human ... CFLAR(8837)
mouse ... Cflar(12633)
rat ... Cflar(117279)

Description

General description

In human Viral FLICE-inhibitory proteins (v-FLIPs) is identified as c-FLIP. It is composed of two death effector domains which have structural resemblance with the N-terminal half of caspase-8 and a caspase-like domain. It exist as multiple splice variants: FLIP α, β, γ and δ. Along with splice variants, it has two endogenous forms of the protein c-FLIPlong and c-FLIPshort.

Specificity

Detects all FLIP splice variants including FLIPα (FLIPL), FLIPβ, FLIPγ, and FLIPδ (FLIPS) by immunoblotting.

Immunogen

synthetic peptide corresponding to amino acids 2-18 of the N-terminal region of FLICE-inhibitory protein (FLIP). This sequence is identical in all FLIP splice variants.

Application

Anti-FLIPL C-Terminal antibody is suitable for immunoblots assay to study the expression levels of elevated c-FLIP protein expression levels in rat carotid arteries five days post-balloon injury. It is also suitable for western blot at a concentration of 0.5-1μg/mL.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

C-FLIP plays an important role in apoptosis signaling pathways. It acts as proapoptotic molecule or as an anti-apoptotic molecule. It has been reported that c-FLIP can interact with both FADD and caspase-8. It prevents caspase-8 recruitment and processing through DED-DED interaction followed by CD95-induced apoptosis.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?