• USA Home
  • FTG224 - FLAG-tag Gene human

EMAIL THIS PAGE TO A FRIEND
FTG224 Sigma-Aldrich

FLAG-tag Gene human

MYF5

Purchase

Properties

description   supplied in Escherichia coli DH5α cells
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... MYF5(4617)

Description

General description

3xFLAG tagged genes are based on end-sequenced fosmid clones, not cDNA. Each tagged clone carries the complete gene of interest embedded in approximately 40 kb of native human or mouse genomic DNA, with the reporter sequence (3xFLAG tag) fused to the extreme 3′-end of the protein-coding sequence.

Application

The 3xFLAG system is an improvement upon the original system by fusing 3 tandem FLAG epitopes (22 amino acids). Detection of fusion proteins containing 3xFLAG is enhanced up to 200 times more than other systems. Like the original FLAG tag, 3xFLAG is hydrophilic, contains an EK cleavage site, and is relatively small. Therefore, the risk of altering protein function, blocking other epitopes or decreasing solubility is minimized.

Legal Information

3xFLAG is a trademark of Sigma-Aldrich Co. LLC

FLAG is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?