EMAIL THIS PAGE TO A FRIEND
G1036 Sigma-Aldrich

Gossypin

≥90% (TLC), from Hibiscus vitifolius

Synonym: 2-(3,4-Dihydroxyphenyl)-8-(β-D-glucopyranosyloxy)-3,5,7-trihydroxy- 4H-1-benzopyran-4-one, 3,3′,4′,5,7,8-Hexahydroxyflavone 8-glucoside, Gossypetin 8-glucoside

  • CAS Number 652-78-8

  • Empirical Formula (Hill Notation) C21H20O13

  • Molecular Weight 480.38

  •  MDL number MFCD00017429

Purchase

Properties

biological source   Hibiscus vitifolius
assay   ≥90% (TLC)
SMILES string   OC[C@H]1O[C@@H](Oc2c(O)cc(O)c3C(=O)C(O)=C(Oc23)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O
InChI   1S/C21H20O13/c22-5-11-13(27)15(29)17(31)21(32-11)34-19-10(26)4-9(25)12-14(28)16(30)18(33-20(12)19)6-1-2-7(23)8(24)3-6/h1-4,11,13,15,17,21-27,29-31H,5H2/t11-,13-,15+,17-,21+/m1/s1
InChI key   SJRXVLUZMMDCNG-KKPQBLLMSA-N

Description

Biochem/physiol Actions

Gossypin is an anti-inflammatory pentahydroxyflavone glucoside isolated from Hibiscus vitifolius that is used as an herbal remedy for diabetes, jaundice, and inflammation. Gossypin potently inhibits human cancer cell proliferation, including melanoma and glioma cells. Gossypin directly binds and inhibits BRAFV600E and CDK4 kinases, and also regulates Chk1 and Cdc25C in U251 glioma cells.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DJ3009900

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Dietary Antioxidants

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.1 During the oxi...
Biofiles Volume 2 Article 2
Keywords: Adhesion, Anti-inflammatory agents, Applications, Cancer, Cardiovascular, Catalysis, Diabetes, Diseases, Gene expression, Metabolism, Neurodegenerative Diseases, Peroxidations, Phosphorylations, Prebiotics, Probiotics, Reductions, Transcription, Transduction, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?