• USA Home
  • G1153 - Glu-Ser-Pro-Leu-Ile-Ala-Lys-Val-Leu-Thr-Thr-Glu-Pro-Pro-Ile-Ile-Thr-Pro-Val-Arg-Arg

EMAIL THIS PAGE TO A FRIEND
G1153 Sigma-Aldrich

Glu-Ser-Pro-Leu-Ile-Ala-Lys-Val-Leu-Thr-Thr-Glu-Pro-Pro-Ile-Ile-Thr-Pro-Val-Arg-Arg

≥95% (HPLC)

Synonym: PLAP, Phospholipase A2 activating peptide

  • CAS Number 137314-60-4

  • Empirical Formula (Hill Notation) C106H184N28O30

  • Molecular Weight 2330.76

  •  MDL number MFCD00171646

Purchase

Properties

assay   ≥95% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C
SMILES string   CC[C@H](C)[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CO)NC(=O)[C@@H](N)CCC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCC(O)=O)C(=O)N2CCC[C@H]2C(=O)N3CCC[C@H]3C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H]([C@@H](C)O)C(=O)N4CCC[C@H]4C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

Description

Biochem/physiol Actions

Induced in smooth muscle and endothelial cells treated with leukotriene D4; activates phospholipase A2.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?