• USA Home
  • G212080 - DMT-2′O-TBDMS-rG(tac) Phosphoramidite

EMAIL THIS PAGE TO A FRIEND
G212080 Sigma-Aldrich

DMT-2′O-TBDMS-rG(tac) Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym: DMT-2′-O-TBDMS-rG(tac) amidite, N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-guanosine,3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite]

  • CAS Number 149989-68-4

  • Empirical Formula (Hill Notation) C58H76N7O10PSi

  • Molecular Weight 1090.32

  •  MDL number MFCD09842116

  •  PubChem Substance ID 329799887

  •  NACRES NA.51

Purchase

Properties

Related Categories 2′O-TBDMS RNA Phosphoramidites, Chemical Synthesis, DMT-2′O-TBDMS-rG(tac) Phosphoramidite, Expedite, Fast Deprotection 2′O-TBDMS RNA Phosphoramidites,
Quality Level   200
product line   Proligo Reagents
assay   ≥96.0% (reversed phase HPLC)
  ≥98% (31P-NMR)
application(s)   oligo synthesis: suitable
λ   conforms (UV/VIS Identity)
compatibility   configured for PerkinElmer
  configured for Polygen
nucleoside profile   base: guanosine
base protecting group: TAC
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: fast
storage temp.   −20°C
SMILES string   COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)COc5ccc(cc5)C(C)(C)C)=Nc34)(c6ccccc6)c7ccc(OC)cc7
InChI   1S/C58H76N7O10PSi/c1-38(2)65(39(3)4)76(72-34-18-33-59)74-50-47(35-71-58(41-19-16-15-17-20-41,42-23-27-44(68-11)28-24-42)43-25-29-45(69-12)30-26-43)73-54(51(50)75-77(13,14)57(8,9)10)64-37-60-49-52(64)62-55(63-53(49)67)61-48(66)36-70-46-31-21-40(22-32-46)56(5,6)7/h15-17,19-32,37-39,47,50-51,54H,18,34-36H2,1-14H3,(H2,61,62,63,66,67)/t47-,50-,51-,54-,76?/m1/s1
InChI key   ODCBQUOWLVQKQG-PZUNXSRFSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?