EMAIL THIS PAGE TO A FRIEND
G2550 Sigma-Aldrich

Sephadex® G-25

Superfine

Purchase

Description

Application

Fractionation Range (MW)
Globular Proteins: 1,000 - 5,000
Dextrans: 100 - 5,000

Legal Information

Sephadex is a registered trademark of GE Healthcare

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Combinatorial synthesis, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Ion exchange resins, Pharmaceutical, Purification, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?