EMAIL THIS PAGE TO A FRIEND
G2795 Sigma

ω-Grammotoxin SIA

from Grammostola spatulata venom, ≥98% (HPLC)

  • CAS Number 152617-90-8

  • Empirical Formula (Hill Notation) C177H268N52O50S6

  • Molecular Weight 4116.73

  •  MDL number MFCD03457797

Purchase

Properties

biological source   Grammostola spatulata venom
assay   ≥98% (HPLC)
solubility   aqueous buffer: soluble
storage temp.   −20°C

Description

Amino Acid Sequence

Asp-Cys-Val-Arg-Phe-Trp-Gly-Lys-Cys-Ser-Gln-Thr-Ser-Asp-Cys-Cys-Pro-His-Leu-Ala-Cys-Lys-Ser-Lys-Trp-Pro-Arg-Asn-Ile-Cys-Val-Trp-Asp-Gly-Ser-Val

Biochem/physiol Actions

Potent inhibitor of both Cav2.1 (P-type) and Cav2.2 (N-type) channels

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?