• USA Home
  • G415050 - DMT-dG(dmf)-CPG Column

EMAIL THIS PAGE TO A FRIEND
G415050 Sigma-Aldrich

DMT-dG(dmf)-CPG Column

50 nmol

Purchase

Properties

Related Categories ABI 39X, Chemical Synthesis, DNA Column, Expedite, Phosphoramidites and Reagents for Oligonucleotide Synthesis (Proligo Reagents),
product line   Proligo Reagents
pore size   1000 Å
compatibility   configured for ABI
  configured for PerkinElmer
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?