• USA Home
  • G435060 - DMT-dG(dmf)-CPG Column
EMAIL THIS PAGE TO A FRIEND
G435060 Sigma-Aldrich
DMT-dG(dmf)-CPG Column
pore size 1000 Å, 1 μmol, configured for ABI, configured for PerkinElmer 89er series
Purchase
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?