• USA Home
  • G5646 - Gly-Arg-Gly-Asp-Thr-Pro

EMAIL THIS PAGE TO A FRIEND
G5646 Sigma-Aldrich

Gly-Arg-Gly-Asp-Thr-Pro

≥97% (HPLC)

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, Cell Signaling and Neuroscience,
biological source   synthetic (organic)
assay   ≥97% (HPLC)
form   powder
storage temp.   −20°C
SMILES string   CC(O)C(NC(=O)C(CC(O)=O)NC(=O)CNC(=O)C(CCCNC(N)=N)NC(=O)CN)C(=O)N1CCCC1C(O)=O
InChI   1S/C23H39N9O10/c1-11(33)18(21(40)32-7-3-5-14(32)22(41)42)31-20(39)13(8-17(36)37)30-16(35)10-28-19(38)12(29-15(34)9-24)4-2-6-27-23(25)26/h11-14,18,33H,2-10,24H2,1H3,(H,28,38)(H,29,34)(H,30,35)(H,31,39)(H,36,37)(H,41,42)(H4,25,26,27)
InChI key   UMZVBZDHGKJFGQ-UHFFFAOYSA-N
Gene Information   human ... COL1A1(1277), COL1A2(1278), FN1(2335), VTN(7448)
mouse ... COL1A1(12842), COL1A2(12843), FN1(14268), VTN(22370)
rat ... COL1A1(29393), COL1A2(84352), FN1(25661), VTN(29169)

Description

Amino Acid Sequence

Gly-Arg-Gly-Asp-Thr-Pro

Biochem/physiol Actions

Inhibits cell attachment of fibronectin, vitronectin, and Type I collagen.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?