• USA Home
  • G6669 - Monoclonal Anti-Gemin 2 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
G6669 Sigma

Monoclonal Anti-Gemin 2 antibody produced in mouse

clone 2E17, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-SIP1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Gene Regulation, Antibodies for Gene Regulation and Expression,
species reactivity   Xenopus, hamster, human, mouse, rat, bovine, canine, chicken
application(s)   immunocytochemistry: suitable
  immunoprecipitation (IP): suitable
  microarray: suitable
  western blot: 0.5-1 μg/mL using HeLa cell extract
clone   2E17, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt ~32 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... SIP1(8487)
mouse ... Sip1(66603)
rat ... Sip1(84404)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

recombinant human Gemin 2.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Application

Monoclonal Anti-Gemin 2 antibody produced in mouse is suitable for immunocytochemistry, immunoprecipitation and microarray. It is also suitable for western blot analysis at a working concentration of 0.5-1μg/mL using HeLa cell extract. The antibody is important in studying the role of the Gemin protein family in nuclear processes.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

GEMIN2 [gem (nuclear organelle) associated protein 2] encodes a protein that forms a multi-protein complex with the survival of motor neuron (SMN) protein. This complex also consists of several other gemin proteins. Gemin2 interacts directly with the SMN protein while Gemin4 and 6 require Gemin3 and 7 respectively for their interaction. The complex is involved in spliceosomal snRNP assembly in the cytoplasm and pre-mRNA splicing and transcription in the nucleus. Deletion or mutation of SMN gene causes spinal muscular atrophy (SMA) disease that is characterized by degeneration of motor neurons and muscular atrophy.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

G5796 Monoclonal Anti-Gemin 8 antibody produced in mouse, ~1 mg/mL, clone 1F8, purified immunoglobulin, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?