• USA Home
  • G7137 - α-D-Glucose 1,6-bisphosphate tetra(cyclohexylammonium) salt hydrate

EMAIL THIS PAGE TO A FRIEND
G7137 Sigma-Aldrich

α-D-Glucose 1,6-bisphosphate tetra(cyclohexylammonium) salt hydrate

≥95%

Synonym: α-D-Glucose 1,6-diphosphate tetra(cyclohexylammonium) salt hydrate, Glc-1,6-PP.4CHA

  • CAS Number 71662-13-0

  • Empirical Formula (Hill Notation) C6H14O12P2 · 4C6H13N · xH2O

  • Molecular Weight 736.81 (anhydrous basis)

  •  EC Number 275-777-4

  •  MDL number MFCD00167199

Purchase

Properties

InChI Key   DMUBREKUECBHKK-KMWDTYPGSA-N
assay   ≥95%
storage temp.   −20°C

Description

Application

Glucose-1,6-bisphosphate and fructose-1,6-bisphosphate were used as bisphosphorylated glycan standards in mass spectrometry analysis to determine if pyrophosphate ions observed in lipid A from Gram-negative bacteria were artifacts produced by condensation of two phosphate groups in the dissociating ion. α-D-Glucose 1,6-bisphosphate is used in the identification, differentiation and characterization of α-phosphoglucomutase(s), β-phosphoglucomutas(s) and α- and β-hexoglucomutase(s).

Preparation Note

Enzymatically prepared.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number

Certificate of Origin

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

G5875 α-D-Glucose 1,6-bisphosphate tetra(cyclohexylammonium) salt hydrate, ≥95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?