• USA Home
  • G9385 - Guanylate Kinase from porcine brain

EMAIL THIS PAGE TO A FRIEND
G9385 Sigma

Guanylate Kinase from porcine brain

buffered aqueous glycerol solution, ≥10 units/mg protein (modified Warburg-Christian)

Synonym: ATP:[d]GMP phosphotransferase, Deoxyguanylate kinase

Purchase

Properties

Related Categories 2.7.x.x Phosphorus containing groups, 2.x.x.x Transferases, Application Index, Biochemicals and Reagents, Cell Signaling Enzymes,
form   buffered aqueous glycerol solution
foreign activity   Myokinase, may contain trace amount
storage temp.   2-8°C

Description

Unit Definition

One unit will convert 1.0 μmole each of GMP and ATP to GDP and ADP per min at pH 7.5 at 30 °C.

Physical form

Solution in 50% glycerol, 1 mM potassium phosphate, pH approx. 7.0, containing 0.1 mm EDTA

Price and Availability


Your Key to Success

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?