• USA Home
  • G9766 - D-(+)-Gluconic acid δ-lactone

EMAIL THIS PAGE TO A FRIEND
G9766 Sigma-Aldrich

D-(+)-Gluconic acid δ-lactone

BioReagent, suitable for cell culture

Synonym: δ-Gluconolactone, 1,2,3,4,5-Pentahydroxycaproic acid δ-lactone, D-(+)-Dextronic acid δ-lactone

  • CAS Number 90-80-2

  • Empirical Formula (Hill Notation) C6H10O6

  • Molecular Weight 178.14

  •  Beilstein/REAXYS Number 83286

  •  EC Number 202-016-5

  •  MDL number MFCD00006647

  •  PubChem Substance ID 24895381

Purchase

Properties

product line   BioReagent
assay   99-100%
application(s)   cell culture | mammalian: suitable
mp   160 °C (dec.) (lit.)
SMILES string   OC[C@H]1OC(=O)[C@H](O)[C@@H](O)[C@@H]1O
InChI   1S/C6H10O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-5,7-10H,1H2/t2-,3-,4+,5-/m1/s1
InChI key   PHOQVHQSTUBQQK-SQOUGZDYSA-N

Description

Biochem/physiol Actions

Glucono-d-lactone increased the doubling time and activated enzymes involved in the oxidative pentose phosphate pathway of Saccharomyces cerevisiae.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
LZ5184000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?