• USA Home
  • GE17-0885-04 - Gammabind g Sepharose®

EMAIL THIS PAGE TO A FRIEND
GE17-0885-04

Gammabind g Sepharose®

Cytiva, 17-0885-04, pack of 250 mL

  •  NACRES NA.56

Purchase

Properties

shelf life   Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
packaging   pack of 250 mL
mfr. no.   Cytiva, 17-0885-04
matrix   4% agarose
particle size   45-165 μm
average diameter   90 μm
cleaning in place   2 - 9 (pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.)
working range   3 - 9
capacity   >17 mg binding capacity(human IgG/ml)
storage temp.   2-8°C

Description

General description

Recombinant GammaBind is an affinity medium for laboratory scale antibody purification. Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse.

Application

GammaBind G Sepharose® is GammaBind G, Type 2, covalently immobilized to Sepharose® 4B by the cyanogen bromide method. GammaBind G, Type 2, a recombinant form of streptococcal Protein G, binds to the Fc region of IgG from a variety of mammalian species. GammaBind G Sepharose® may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium

Features and Benefits

• Recombinant GammaBind G type 2 coupled to Sepharose® 4B engineered for antibody purification at laboratory scale.
• Binds to all IgG subclasses from human, mouse, and rat.
• Binds total IgG from guinea pig, rabbit, goat, cow sheep, and horse.
• Will not cross-react with other serum Proteins such as IgM, IgE, IgA or transferrin.

Analysis Note

www.gelifesciences.com.

Legal Information

Sepharose is a registered trademark of Cytiva

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?