• USA Home
  • GW21186 - Anti-CDH1 antibody produced in chicken

EMAIL THIS PAGE TO A FRIEND
GW21186 Sigma-Aldrich

Anti-CDH1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-Epithelial-cadherin, Anti-Cadherin-1

Purchase

Properties

Related Categories A to L, Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Adhesion Molecules and Related Proteins,
conjugate   unconjugated
clone   polyclonal
biological source   chicken
application(s)   western blot: suitable
species reactivity   human, rat, mouse
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
mfr. no.   Genway, 15-288-21186
NCBI accession no.   NP_004351.1
UniProt accession no.   P12830
Gene Information   human ... CDH1(999)

Description

Immunogen

Immunogen Sequence: GI # 4757960, sequence 441-523

Recombinant cadherin 1, type 1 preproprotein

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?