• USA Home
  • GW21948F - Anti-SUGT1 antibody produced in chicken

EMAIL THIS PAGE TO A FRIEND
GW21948F Sigma

Anti-SUGT1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-SGT1, suppressor of G2 allele of SKP1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, IgY Antibodies, M to Z, Primary Antibodies,
species reactivity   human
application(s)   western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... SUGT1(10910)
biological source   chicken
antibody product type   primary antibodies
mfr. no.   Genway, 15-288-21948F
conjugate   unconjugated
NCBI accession no.   NP_006695.1
UniProt accession no.   Q5TAM5

Description

Immunogen

Immunogen Sequence: GI # 5730041, sequence 1-333

Recombinant suppressor of G2 allele of SKP1

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Amplification, Buffers, Diagnostic, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Phosphorylations, Purification, Therapeutic uses, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?