• USA Home
  • GW22005A - Anti-GTF2H1 (ab1) antibody produced in chicken

EMAIL THIS PAGE TO A FRIEND
GW22005A Sigma

Anti-GTF2H1 (ab1) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-TFIIH basal transcription factor complex p62 subunit

Purchase

Properties

Related Categories A to L, Alphabetical Index, Antibodies, Antibodies to Transcription Factors, Epigenetics,
species reactivity   rat, mouse, human
application(s)   western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... GTF2H1(2965)
biological source   chicken
antibody product type   primary antibodies
mfr. no.   Genway, 15-288-22005A
conjugate   unconjugated
NCBI accession no.   NP_005307.1
UniProt accession no.   P32780

Description

Immunogen

Immunogen Sequence: GI # 4885365, sequence 1-219

Recombinant general transcription factor IIH, polypeptide 1 (62kD subunit)

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Therapeutic uses, Western blot

抗体基础知识

免疫球蛋白(Ig)由B淋巴细胞产生并分泌到血浆中。单体形式的Ig分子是一个分子量约为150 kDa的糖蛋白,且其结构差不多类似字母Y。Ig单体的基本结构(图1)包括由两个二硫键连接的两个相同的半体。每个半体都具有一个约50 kDa的重链和一个约25 kDa的轻链,两者在轻链羧基末端附近通过二硫键连接在一起。重链可分为Fc部分和Fab部分,前者位于羧基末端(Y的基座),后者则位于氨基末端(Y的臂)...
Keywords: Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Rheumatology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?