• USA Home
  • GW22333B - Anti-HEYL (ab2) antibody produced in chicken

EMAIL THIS PAGE TO A FRIEND
GW22333B Sigma-Aldrich

Anti-HEYL (ab2) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Purchase

Properties

Related Categories A to L, Alphabetical Index, Antibodies, Antibodies for Epigenetics and Nuclear Signaling, Antibodies to Transcription Factors,
conjugate   unconjugated
clone   polyclonal
biological source   chicken
application(s)   western blot: suitable
species reactivity   human
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
mfr. no.   Genway, 15-288-22333B
NCBI accession no.   NP_055386.1
UniProt accession no.   Q5TG99
Gene Information   human ... HEYL(26508)

Description

General description

HEYL (Hairy/enhancer-of-split related with YRPW motif-like) is a transcriptional repressor belonging to the
family of basic helix-loop-helix-type repressors. It is a protein encoded by the HEYL gene in humans and is mapped to chromosome 1p34.

Immunogen

Immunogen Sequence: GI # 7657154, sequence 231-328

Recombinant hairy/enhancer-of-split related with YRPW motif-like

Application

Anti-HEYL (ab2) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Biochem/physiol Actions

HEYL (Hairy/enhancer-of-split related with YRPW motif-like) protein is a protein encoded by the HEYL gene in humans and is mapped to chromosome 1p34. The protein is a transcriptional repressor and downstream effector of Notch signaling. HEYL acts as a more potent repressor of androgen receptor (AR) activity. It helps in repressing endogenous AR-regulated genes and reduces androgen-dependent prostate cancer cell growth. Nuclear exclusion of HEY proteins may be a critical step in the progression of prostate cancer.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?