• USA Home
  • H141 - S(−)-8-Hydroxy-DPAT hydrobromide

EMAIL THIS PAGE TO A FRIEND
H141 Sigma-Aldrich

S(−)-8-Hydroxy-DPAT hydrobromide

solid

Synonym: S(−)-8-OH-DPAT HBr

  • CAS Number 78950-78-4

  • Empirical Formula (Hill Notation) C16H25NO · HBr

  • Molecular Weight 328.29

  •  MDL number MFCD00153811

  •  PubChem Substance ID 24895459

Purchase

Properties

Related Categories Agonists, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience,
InChI Key   BATPBOZTBNNDLN-UQKRIMTDSA-N
form   solid
optical activity   [α]25/D −67.8° in methanol(lit.)
color   white

Description

Packaging

25, 100 mg in poly bottle

5 mg in glass bottle

Biochem/physiol Actions

Partial 5-HT1A serotonin receptor agonist.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Making Headway

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?