• USA Home
  • H4629 - 12-Hydroxystearyl alcohol

EMAIL THIS PAGE TO A FRIEND
H4629 Sigma-Aldrich

12-Hydroxystearyl alcohol

≥99%

Synonym: 1,12-Octadecanediol

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acids and conjugates, Fatty Acyls, Fatty Alcohols, Hydroxy Fatty Acids,
InChI Key   KHLCTMQBMINUNT-UHFFFAOYSA-N
assay   ≥99%

Description

Packaging

1 g in poly bottle

Preparation Note

Prepared from ricinoleic acid

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?