• USA Home
  • H4649 - (Hydroxypropyl)methyl cellulose

EMAIL THIS PAGE TO A FRIEND
H4649 Sigma

(Hydroxypropyl)methyl cellulose

viscosity ~4000 cP, 2 % in H2O(20 °C)(lit.)

Purchase

Properties

viscosity   ~4000 cP, 2 % in H2O(20 °C)(lit.)

Description

Application

Thickener for aqueous and non-aqueous systems, clear films with grease resistance, binders, lubricants, steric stabilizer and water retention aid.

Features and Benefits

Dissolves in water, undergoes reversible gelation upon heating, non-ionic, does not complex with ionic species and is surface active and enzyme resistant. Solutions are pseudoplastic.

Suitability

Suitable for use as an additive or modifier in capillary electrophoresis to separate proteins, peptides, or nucleic acids.

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?