• USA Home
  • H5377 - 17α-Hydroxypregnenolone 3,17-diacetate

EMAIL THIS PAGE TO A FRIEND
H5377 Sigma-Aldrich

17α-Hydroxypregnenolone 3,17-diacetate

Synonym: 3β,17α-Dihydroxy-5-pregnen-20-one 3,17-diacetate, 5-Pregnene-3β,17α-diol-20-one 3,17-diacetate

  • CAS Number 1176-21-2

  • Empirical Formula (Hill Notation) C25H36O5

  • Molecular Weight 416.55

  •  MDL number MFCD00021128

Purchase

Properties

SMILES string   CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(OC(C)=O)C(C)=O)[C@@H]3CC=C2C1
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?