• USA Home
  • H5771 - DL-α-Hydroxyarachidic acid

EMAIL THIS PAGE TO A FRIEND
H5771 Sigma-Aldrich

DL-α-Hydroxyarachidic acid

≥98% (capillary GC)

Synonym: 2-Hydroxyeicosanoic acid

  • CAS Number 16742-48-6

  • Empirical Formula (Hill Notation) C20H40O3

  • Molecular Weight 328.53

  •  MDL number MFCD00083367

Purchase

Properties

assay   ≥98% (capillary GC)
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCCCCCCCC(O)C(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?