• USA Home
  • H5779 - Hemocyanin-Agarose from Megathura crenulata (keyhole limpet)

EMAIL THIS PAGE TO A FRIEND
H5779 Sigma-Aldrich

Hemocyanin-Agarose from Megathura crenulata (keyhole limpet)

PBS suspension

Synonym: KLH-Agarose

Purchase

Properties

form   PBS suspension
extent of labeling   3-4 mg per mL
matrix   4% beaded agarose
matrix activation   cyanogen bromide
storage temp.   2-8°C

Description

Physical form

Suspension in 0.01 M phosphate buffered saline, pH 7.4, containing 0.1% sodium azide

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?