• USA Home
  • H6896 - Methyl 2-hydroxytetradecanoate

EMAIL THIS PAGE TO A FRIEND
H6896 Sigma-Aldrich

Methyl 2-hydroxytetradecanoate

≥98% (capillary GC)

Synonym: DL-α-Hydroxymyristic acid methyl ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acyls, Fatty Esters, Hydroxy fatty acids, Lipids,
assay   ≥98% (capillary GC)
form   solid
functional group   ester
shipped in   ambient
storage temp.   2-8°C
SMILES string   CCCCCCCCCCCCC(O)C(=O)OC
InChI   1S/C15H30O3/c1-3-4-5-6-7-8-9-10-11-12-13-14(16)15(17)18-2/h14,16H,3-13H2,1-2H3
InChI key   ATFSJSYRBAGGIS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SPB®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SPB®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?