• USA Home
  • HPA003887 - Anti-LAX1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
HPA003887 Sigma-Aldrich

Anti-LAX1 antibody produced in rabbit

Antibody Enhanced Validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-Membrane-associated adapter protein LAX antibody produced in rabbit, Anti-Linker for activation of X cells antibody produced in rabbit, Anti-Lymphocyte transmembrane adapter 1 antibody produced in rabbit

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, L, L-LL, Prestige Antibodies,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunofluorescence: 1-4 μg/mL
  immunohistochemistry: 1:500- 1:1000
species reactivity   human
enhanced validation   orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
grade   Prestige Antibodies® Powered by Atlas Antibodies
immunogen sequence   PWRQEDLGRHESRSMRIFSTESLLSRNSESPEHVPSQAGNAFQEHTAHIHATEYAVGIYDNAMVPQMCGNLTPSAHCINVRASRDCASISSEDSHDYVNVPTAEEIAETLASTKSPSRNLFVLPSTQKLEFTEERDEGCGDAGDC
UniProt accession no.   Q8IWV1
Gene Information   human ... LAX1(54900)

Description

Immunogen

Lymphocyte transmembrane adapter 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project (www.proteinatlas.org)and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Sigma-Aldrich Co. LLC

Biochem/physiol Actions

Lymphocyte transmembrane adaptor 1 (LAX1) is a protein referred to as LAX. It is mainly expressed in B cells, T cell and other lymphoid-specific cell types. It negatively regulates signaling in lymphocytes. It is a novel type of membrane microdomain containing a number of physiologically important proteins. The inhibition of signaling events involved in T cell activation by this gene occurs through both dependent on and independent mechanism of its tyrosine phosphorylation. The protein binds by its N terminus to active GTP-Rab8, as well as the cytoplasmic tail of CTLA-4. It acts as an effector complex in the transport of CTLA-4 to the surfaces of T cells. It is a linker for activation of T cells (LAT)-like molecule that is expressed in lymphoid tissues.

Linkage

Corresponding Antigen APREST86508.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Prestige Antibody® Immunofluorescence Procedure

Prestige Antibodies® have been used for subcellular localization studies by immunocytochemistry (ICC) - immunofluorescence (IF) using the protocol described below. In addition to the primary antibodi...
Keywords: Immunocytochemistry, Immunofluorescence, Immunostaining, Sample preparations

Prestige Antibody® Immunohistochemistry Procedure

Prestige Antibodies® developed for immunohistochemistry based expression profiling are recommended to be used according to the standard IHC staining protocol described below.
Keywords: Buffers, Dehydration reaction, Gene expression, Hydration reaction, Immunohistochemistry, Immunostaining, Tissue microarrays

Prestige Antibody® Protein Immunoblotting Procedure

Prestige Antibodies® recommended for Western Blot are recommended to be used according to the standard WB protocol described below. Western blot standard protocol optimized for
Keywords: Catalytic combustion detector, Electrophoresis, Gene expression, Sample preparations, Western blot

Prestige Antigens™-Blocking Protocol

Prestige Antigens and the corresponding Prestige Antibodies are recommended to be used according to the standard blocking protocol described below.
Keywords: Immunocytochemistry, Immunohistochemistry, Immunostaining, Titrations, Western blot

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?