• USA Home
  • HPA011220 - Anti-PCDH10 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
HPA011220 Sigma-Aldrich

Anti-PCDH10 antibody produced in rabbit

Antibody Enhanced Validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-Protocadherin-10 precursor

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, P-PF, PB-PC, Prestige Antibodies,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry: 1:50- 1:200
species reactivity   human
enhanced validation   orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
grade   Prestige Antibodies® Powered by Atlas Antibodies
immunogen sequence   KEKKLNIYTCLASDCCLCCCCCGGGGSTCCGRQARARKKKLSKSDIMLVQSSNVPSNPAQVPIEESGGFGSHHHNQNYCYQVCLTPESAKTDLMFLKPCSPSRSTDTEHNPCGAIVTGYTDQQPDIISNGSILSNETKHQRA
UniProt accession no.   Q9P2E7
Gene Information   human ... PCDH10(57575)

Description

General description

PCDH10 (protocadherin 10) belongs to the cadherin superfamily and is a potential tumor suppressor gene. This gene maps to human chromosome 4q28.3, and has wide range of adult tissue expression. It is a part of the protocadherin subfamily where it belongs to the δ2 subgroup.

Immunogen

Protocadherin-10 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PCDH10 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project (www.proteinatlas.org). Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Legal Information

Prestige Antibodies is a registered trademark of Sigma-Aldrich Co. LLC

Linkage

Corresponding Antigen APREST71992.

Biochem/physiol Actions

PCDH10 (protocadherin 10) is a tumor suppressor gene which is inactivated epigenetically in various human cancers. In colorectal cancer, the promoter of this gene is methylated leading to suppression of PCDH10 expression. It prevents angiogenesis in tumors, and suppresses multiple myeloma. It is involved in the cell cycle control and proliferation of multiple myeloma. It is thought to be involved in the determination and function of certain cell-cell connections in the brain. PCDH10 is epigenetically and genetically under-expressed in medulloblastoma patients, and this down-regulation might be responsible for tumor progression and metastasis. It also has p53 binding site in its promoter region, and its expression is induced by p53, which is a tumor suppressor gene. Methylation of CpG sites of PCDH10 promoter is associated with poor prognosis of gastric cancer patients.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Prestige Antibody® Immunofluorescence Procedure

Prestige Antibodies® have been used for subcellular localization studies by immunocytochemistry (ICC) - immunofluorescence (IF) using the protocol described below. In addition to the primary antibodi...
Keywords: Immunocytochemistry, Immunofluorescence, Immunostaining, Sample preparations

Prestige Antibody® Immunohistochemistry Procedure

Prestige Antibodies® developed for immunohistochemistry based expression profiling are recommended to be used according to the standard IHC staining protocol described below.
Keywords: Buffers, Dehydration reaction, Gene expression, Hydration reaction, Immunohistochemistry, Immunostaining, Tissue microarrays

Prestige Antibody® Protein Immunoblotting Procedure

Prestige Antibodies® recommended for Western Blot are recommended to be used according to the standard WB protocol described below. Western blot standard protocol optimized for
Keywords: Catalytic combustion detector, Electrophoresis, Gene expression, Sample preparations, Western blot

Prestige Antigens™-Blocking Protocol

Prestige Antigens and the corresponding Prestige Antibodies are recommended to be used according to the standard blocking protocol described below.
Keywords: Immunocytochemistry, Immunohistochemistry, Immunostaining, Titrations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?