• USA Home
  • HPA026827 - Anti-MUL1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
HPA026827 Sigma-Aldrich

Anti-MUL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-C1orf166, Anti-FLJ12875, Anti-GIDE, Anti-MAPL, Anti-MULAN, Anti-RNF218, Anti-mitochondrial E3 ubiquitin ligase 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, ML-MZ, MUE-MZ, Prestige Antibodies,
product line   Prestige Antibodies® Powered by Atlas Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  indirect immunofluorescence: suitable
  western blot: suitable
species reactivity   human
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
immunogen sequence   EFQEHEAQLLSRAKPEDRESLKSACVVCLSSFKSCVFLECGHVCSCTECYRALPEPKKCPI
UniProt accession no.   Q969V5
Gene Information   human ... MUL1(79594)

Description

General description

The gene MUL1 (mitochondrial ubiquitin ligase activator of NFκB1) is mapped to human chromosome 1p36.12. The encoded protein has two transmembrane domain and a conserved C-terminal RING (really interesting new gene)-finger domain. It is attached to the mitochondrial outer membrane. MUL1 is also referred to as Hades or MULAN (mitochondrial ubiquitin ligase activator of NFκB1) or GIDE (growth inhibition and death E3 ligase).

Immunogen

mitochondrial E3 ubiquitin ligase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project (www.proteinatlas.org). Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Anti-MUL1 antibody produced in rabbit has been used in western blotting.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Legal Information

Prestige Antibodies is a registered trademark of Sigma-Aldrich Co. LLC

Biochem/physiol Actions

MUL1 (mitochondrial ubiquitin ligase activator of NFκB1) controls various cellular proteins, such as NF-κB (nuclear factor κ B), JNK (Jun amino-terminal kinase), Akt (serine/threonine protein kinase), GABARAP (γ-aminobutyric acid receptor-associated protein ) and DRP1 (dynamin-related protein 1), thereby regulating cellular signaling. It has role in cell death, proliferation, mitophagy, mitochondrial dynamics and antiviral mechanism. MUL1 also plays a crucial role in ubiquitination of p53 and blocks the interaction between p53 and BCL2 (B-cell lymphoma 2). The protein suppresses p53-dependent mitochondrial cell death pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
• IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
• Protein array of 364 human recombinant protein fragments.
• Validated for multiple commonly used applications such as IHC (Immunohistochemistry), IF (Immunofluorescence), and WB (Western Blot)

Linkage

Corresponding Antigen APREST72832.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Prestige Antibody® Immunofluorescence Procedure

Prestige Antibodies® have been used for subcellular localization studies by immunocytochemistry (ICC) - immunofluorescence (IF) using the protocol described below. In addition to the primary antibodi...
Keywords: Immunocytochemistry, Immunofluorescence, Immunostaining, Sample preparations

Prestige Antibody® Immunohistochemistry Procedure

Prestige Antibodies® developed for immunohistochemistry based expression profiling are recommended to be used according to the standard IHC staining protocol described below.
Keywords: Buffers, Dehydration reaction, Gene expression, Hydration reaction, Immunohistochemistry, Immunostaining, Tissue microarrays

Prestige Antibody® Protein Immunoblotting Procedure

Prestige Antibodies® recommended for Western Blot are recommended to be used according to the standard WB protocol described below. Western blot standard protocol optimized for
Keywords: Catalytic combustion detector, Electrophoresis, Gene expression, Sample preparations, Western blot

Prestige Antigens™-Blocking Protocol

Prestige Antigens and the corresponding Prestige Antibodies are recommended to be used according to the standard blocking protocol described below.
Keywords: Immunocytochemistry, Immunohistochemistry, Immunostaining, Titrations, Western blot

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?