• USA Home
  • HPA040731 - Anti-IFT88 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
HPA040731 Sigma-Aldrich

Anti-IFT88 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-D13S1056E, Anti-MGC26259, Anti-TTC10, Anti-Tg737, Anti-hTg737, Anti-intraflagellar transport 88 homolog (Chlamydomonas)

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, I-IG, I-J, Prestige Antibodies,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
species reactivity   human
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
grade   Prestige Antibodies® Powered by Atlas Antibodies
immunogen sequence   YKEALRNDSSCTEALYNIGLTYEKLNRLDEALDCFLKLHAILRNSAEVLYQIANIYELMENPSQAIEWLMQVVSVIPTDPQVLSKLGELYD
UniProt accession no.   Q13099
Gene Information   human ... IFT88(8100)

Description

Immunogen

intraflagellar transport 88 homolog (Chlamydomonas) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project (www.proteinatlas.org)and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Legal Information

Prestige Antibodies is a registered trademark of Sigma-Aldrich Co. LLC

Linkage

Corresponding Antigen APREST81284.

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Prestige Antibody® Immunofluorescence Procedure

Prestige Antibodies® have been used for subcellular localization studies by immunocytochemistry (ICC) - immunofluorescence (IF) using the protocol described below. In addition to the primary antibodi...
Keywords: Immunocytochemistry, Immunofluorescence, Immunostaining, Sample preparations

Prestige Antibody® Immunohistochemistry Procedure

Prestige Antibodies® developed for immunohistochemistry based expression profiling are recommended to be used according to the standard IHC staining protocol described below.
Keywords: Buffers, Dehydration reaction, Gene expression, Hydration reaction, Immunohistochemistry, Immunostaining, Tissue microarrays

Prestige Antibody® Protein Immunoblotting Procedure

Prestige Antibodies® recommended for Western Blot are recommended to be used according to the standard WB protocol described below. Western blot standard protocol optimized for
Keywords: Catalytic combustion detector, Electrophoresis, Gene expression, Sample preparations, Western blot

Prestige Antigens™-Blocking Protocol

Prestige Antigens and the corresponding Prestige Antibodies are recommended to be used according to the standard blocking protocol described below.
Keywords: Immunocytochemistry, Immunohistochemistry, Immunostaining, Titrations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?