• USA Home
  • I1529 - Anti-IRAK2, C-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
I1529 Sigma-Aldrich

Anti-IRAK2, C-Terminal antibody produced in rabbit

~0.5 mg/mL, IgG fraction of antiserum

Synonym: Anti-Interleukin-1 Receptor-Associated Kinase 2

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Gene Regulation and Expression, Antibodies for Kinase/Phosphatase Biology,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 0.5-1.0 μg/mL using K562 cell lysates
species reactivity   human
mol wt   antigen mol wt 65 kDa
shipped in   dry ice
storage temp.   −20°C
antibody form   IgG fraction of antiserum
antibody product type   primary antibodies
concentration   ~0.5 mg/mL
UniProt accession no.   O43187
Gene Information   human ... IRAK2(3656)

Description

General description

The gene IRAK2 (interleukin 1 receptor associated kinase 2) encodes a member of the IRAK/Pelle family. It is an important signaling molecule of the TLR (Toll like receptor)/IL1-R (interleukin 1 receptor) superfamily. The members of IRAK family are characterized by the presence of an N-terminal DD (death domain) and a C-terminal Ser/Thr kinase or kinase-like domain. The gene is mapped to human chromosome 3p25.3-3p24.1.

Immunogen

synthetic peptide corresponding to amino acids 571-590 of the C-terminal of human IL-1 receptor-associated kinase 2 (IRAK2).

Biochem/physiol Actions

The members of this family activate the transcription factor NF-κB (nuclear factor κ B) via IL-R1 (interleukin receptor type 1) and TLR (Toll-like receptor) mediated inflammatory/immune response. The receptor IL-R1 binds to its ligand proinflammatory cytokine IL-1, and this binding facilitates the association of IL-R1 with IL-1 receptor accessory protein (IL-1RAcP). This complex then recruits myeloid differentiation protein (MyD88), an intracellular TIR-containing adaptor. MyD88, then recruits IRAK1, IRAK2, IRAK4 and IRAK-M. These kinases interact with TRAF6 (TNF receptor-associated factor 6), which in turn links to NF-κB-inducing kinase (NIK). NIK activates the IκB kinase complex (IKKα and IKKβ), which in turn phosphorylates IκB. This final phosphorylation leads to ubiquitin-proteasome-mediated degradation resulting in the activation of NF-κB.

Safety & Documentation

Safety Information

WGK Germany 
-
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

I1654 Anti-IRAK2 (546-564) antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?